Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024403.2 | NP_077379.1 | 3 | 780 | ||
GenBank mRNA | AF252627.1 | - | 2 | 546 | ||
BC158588.1 | - | 3 | 753 | 9098368 | ||
FQ210065.1 | - | 3 | 780 | |||
FQ213021.1 | - | 3 | 780 | |||
FQ214151.1 | - | 3 | 779 | |||
FQ214217.1 | - | 3 | 782 | |||
FQ215912.1 | - | 3 | 778 | |||
FQ219043.1 | - | 3 | 780 | |||
FQ221881.1 | - | 3 | 780 | |||
FQ230257.1 | - | 2 | 408 | |||
FQ232693.1 | - | 3 | 786 |