Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024153.1 | NP_077067.1 | 8, 9 | 828 | ||
GenBank mRNA | BC088844.1 | - | 8, 9 | 835 | 7308634 | |
D63761.1 | - | 8, 9 | 828 |