Gene Symbol: Serpind1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024382.1 | NP_077358.1 | 3, 4 | 956 | ||
XM_006248709.2 | XP_006248771.1 | 2, 3 | 963 | |||
GenBank mRNA | BC091145.1 | - | 3, 4 | 936 | 7372906 | |
FQ218607.1 | - | 2, 3 | 933 | |||
X74549.1 | - | 3, 4 | 956 | |||
X74550.1 | - | 2, 3 | 903 |