Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adamts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024400.2 | NP_077376.2 | 6 | 2121 | ||
GenBank mRNA | AF149118.1 | - | 6 | 2134 | ||
AF304446.1 | - | 6 | 1692 | |||
BC080237.1 | - | 6 | 2121 | 7100443 |