Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031002.3 | NP_112264.3 | 5, 6 | 564 | ||
GenBank mRNA | U26397.1 | - | 5, 6 | 564 |