Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Stx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031124.1 | NP_112386.1 | 3 | 205 | ||
XM_006231097.2 | XP_006231159.1 | 3 | 289 | |||
XM_008760262.1 | XP_008758484.1 | 3 | 289 | |||
XM_008760263.1 | XP_008758485.1 | 3 | 289 | |||
XM_008760264.1 | XP_008758486.1 | 3 | 289 | |||
XM_008760265.1 | XP_008758487.1 | 3 | 289 | |||
XM_008760266.1 | XP_008758488.1 | 3 | 289 | |||
XM_008760267.1 | XP_008758489.1 | 3 | 289 | |||
GenBank mRNA | BC168246.1 | - | 3 | 159 | 5600098 | |
L20820.1 | - | 3 | 205 |