Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017960.1 | NP_001017960.1 | 3 | 525 | ||
NM_031134.2 | NP_112396.2 | 2 | 393 | |||
GenBank mRNA | CB580585.1 | - | 2 | 325 | ||
M18028.1 | - | 3 | 415 | |||
M31174.1 | - | 607 | ||||
X07409.1 | - | 1 | 54 | |||
X12744.1 | - | 2 | 57 |