Search Thermo Fisher Scientific
Gene Symbol: Stc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031123.2 | NP_112385.1 | 3 | 583 | ||
GenBank mRNA | BC078803.1 | - | 3 | 583 | 7123665 | |
U62667.1 | - | 3 | 435 |