Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynlt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031318.1 | NP_112608.1 | 2, 3 | 91 | ||
XM_006227901.2 | XP_006227963.1 | 2, 3 | 156 | |||
GenBank mRNA | AB010119.1 | - | 2, 3 | 80 | ||
AJ131437.1 | - | 2, 3 | 91 | |||
BC166879.1 | - | 2, 3 | 123 | 6919674 |