Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031119.2 | NP_112381.2 | 5 | 513 | ||
XM_006234321.2 | XP_006234383.1 | 5 | 589 | |||
XM_006234322.2 | XP_006234384.1 | 5 | 589 | |||
XM_006234323.2 | XP_006234385.1 | 5 | 589 | |||
GenBank mRNA | BC081780.1 | - | 5 | 513 | 7192454 | |
BC097252.1 | - | 5 | 519 | |||
X67859.1 | - | 4 | 448 |