Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031235.1 | NP_112514.1 | 4, 5 | 845 | ||
XM_006255828.1 | XP_006255890.1 | 4, 5 | 830 | |||
XM_006255829.1 | XP_006255891.1 | 4, 5 | 830 | |||
XM_006255830.1 | XP_006255892.1 | 4, 5 | 830 | |||
XM_006255831.1 | XP_006255893.1 | 4, 5 | 830 | |||
XM_006255832.1 | XP_006255894.1 | 4, 5 | 830 | |||
XM_006255833.1 | XP_006255895.1 | 4, 5 | 830 | |||
XM_006255835.1 | XP_006255897.1 | 4, 5 | 830 | |||
XM_006255836.1 | XP_006255898.1 | 4, 5 | 830 | |||
XM_006255837.1 | XP_006255899.1 | 4, 5 | 830 | |||
XM_006255838.1 | XP_006255900.1 | 4, 5 | 830 | |||
XM_006255839.1 | XP_006255901.1 | 4, 5 | 830 | |||
GenBank mRNA | AB005549.1 | - | 4, 5 | 845 |