Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Gsta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031509.2 | NP_113697.1 | 3, 4 | 375 | ||
GenBank mRNA | AF111160.1 | - | 3, 4 | 375 | ||
AF127460.1 | - | 2, 3 | 145 | |||
BC059128.1 | - | 3, 4 | 191 | 6922234 | ||
BC088127.1 | - | 3, 4 | 175 | 7322480 | ||
FJ179398.1 | - | 2, 3 | 136 | |||
FQ209644.1 | - | 3, 4 | 191 | |||
FQ211002.1 | - | 3, 4 | 186 | |||
FQ211022.1 | - | 3, 4 | 187 | |||
FQ213952.1 | - | 3, 4 | 326 | |||
FQ218347.1 | - | 3, 4 | 187 | |||
FQ218448.1 | - | 3, 4 | 184 | |||
FQ218456.1 | - | 3, 4 | 187 | |||
FQ218470.1 | - | 3, 4 | 189 | |||
FQ218508.1 | - | 3, 4 | 186 | |||
FQ219165.1 | - | 3, 4 | 187 | |||
FQ219196.1 | - | 3, 4 | 187 | |||
FQ219451.1 | - | 3, 4 | 195 | |||
FQ219713.1 | - | 3, 4 | 187 | |||
FQ226984.1 | - | 3, 4 | 189 | |||
FQ229239.1 | - | 3, 4 | 187 | |||
K01932.1 | - | 3, 4 | 180 | |||
S72505.1 | - | 3, 4 | 285 | |||
X78848.1 | - | 3, 4 | 285 |
Gene Symbol: LOC102550391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244659.2 | XP_006244721.2 | 370 | |||
GenBank mRNA | FQ218982.1 | - | 188 |