Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pdgfrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031525.1 | NP_113713.1 | 4 | 770 | ||
XM_006254789.2 | XP_006254851.1 | 4 | 860 | |||
XM_008772084.1 | XP_008770306.1 | 3 | 411 | |||
GenBank mRNA | AY090783.1 | - | 4 | 770 | ||
Z14119.1 | - | 2 | 303 |