Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Add3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164103.1 | NP_001157575.1 | 2 | 292 | ||
NM_031552.2 | NP_113740.2 | 2 | 292 | |||
XM_006231599.2 | XP_006231661.1 | 2 | 145 | |||
XM_006231600.2 | XP_006231662.1 | 3 | 392 | |||
XM_006231601.2 | XP_006231663.1 | 2 | 991 | |||
XM_006231602.2 | XP_006231664.1 | 2 | 993 | |||
XM_008760510.1 | XP_008758732.1 | 2 | 135 | |||
XM_008760511.1 | XP_008758733.1 | 2 | 990 | |||
GenBank mRNA | CB757924.1 | - | 2 | 343 | ||
FM038946.1 | - | 2 | 361 | |||
FM072500.1 | - | 2 | 127 | |||
U35775.1 | - | 209 |