Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031357.1 | NP_112647.1 | 4 | 364 | ||
GenBank mRNA | AB043870.1 | - | 4 | 364 | ||
AY724484.1 | - | 4 | 365 | |||
BC081775.1 | - | 4 | 373 | 7107183 |