Search Thermo Fisher Scientific
Gene Symbol: LOC24906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031537.1 | NP_113725.1 | 4 | 341 | ||
XM_006246376.2 | XP_006246438.1 | 4 | 391 | |||
XM_006246377.2 | XP_006246439.1 | 4 | 421 | |||
GenBank mRNA | AF041083.1 | - | 4 | 341 |