Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031583.2 | NP_113771.2 | 6 | 412 | ||
GenBank mRNA | EV773232.1 | - | 3 | 176 | ||
FQ231827.1 | - | 5 | 249 | |||
FQ231919.1 | - | 6 | 424 | |||
FQ233317.1 | - | 6 | 424 | |||
FQ234437.1 | - | 6 | 424 | |||
U82626.1 | - | 6 | 379 |