Gene Symbol: Gsta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031509.2 | NP_113697.1 | 5 | 601 | ||
GenBank mRNA | AF111160.1 | - | 5 | 601 | ||
AF127460.1 | - | 4 | 371 | |||
BC059128.1 | - | 5 | 417 | 6922234 | ||
BC088127.1 | - | 5 | 401 | 7322480 | ||
FJ179398.1 | - | 4 | 362 | |||
FQ209644.1 | - | 5 | 417 | |||
FQ211002.1 | - | 5 | 412 | |||
FQ211022.1 | - | 5 | 413 | |||
FQ213952.1 | - | 5 | 552 | |||
FQ218347.1 | - | 5 | 413 | |||
FQ218448.1 | - | 5 | 410 | |||
FQ218456.1 | - | 5 | 413 | |||
FQ218470.1 | - | 5 | 415 | |||
FQ218508.1 | - | 5 | 412 | |||
FQ219165.1 | - | 5 | 413 | |||
FQ219196.1 | - | 5 | 413 | |||
FQ219451.1 | - | 5 | 421 | |||
FQ219713.1 | - | 5 | 413 | |||
FQ226984.1 | - | 5 | 415 | |||
FQ229239.1 | - | 5 | 413 | |||
K01932.1 | - | 5 | 406 | |||
S72505.1 | - | 5 | 511 | |||
X78848.1 | - | 5 | 511 |
Gene Symbol: LOC102550391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244659.2 | XP_006244721.2 | 596 | |||
GenBank mRNA | FQ218982.1 | - | 414 |