Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ampd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031544.1 | NP_113732.1 | 6 | 1023 | ||
XM_006230010.2 | XP_006230072.1 | 6 | 1791 | |||
XM_006230011.2 | XP_006230073.1 | 6 | 1156 | |||
XM_006230012.2 | XP_006230074.1 | 6 | 1626 | |||
XM_006230013.2 | XP_006230075.1 | 6 | 1638 | |||
XM_006230014.2 | XP_006230076.1 | 6 | 1542 | |||
XM_006230015.2 | XP_006230077.1 | 6 | 1770 | |||
XM_008759688.1 | XP_008757910.1 | 6 | 1033 | |||
GenBank mRNA | U90888.1 | - | 6 | 1023 |