Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031581.2 | NP_113769.1 | 1 | 279 | ||
GenBank mRNA | U42388.1 | - | 1 | 281 |
Set Membership: |
![]() |