Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031648.2 | NP_113836.2 | 3 | 166 | ||
XM_006228838.2 | XP_006228900.1 | 4 | 386 | |||
XM_006228839.2 | XP_006228901.1 | 3 | 161 | |||
XM_008759184.1 | XP_008757406.1 | 3 | 175 | |||
GenBank mRNA | CB712570.1 | - | 3 | 166 | ||
FM109156.1 | - | 3 | 120 | |||
FQ211349.1 | - | 3 | 177 | |||
FQ224131.1 | - | 3 | 129 | |||
FQ224338.1 | - | 3 | 212 | |||
U72246.1 | - | 2 | 80 |