Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031695.2 | NP_113883.2 | 3 | 643 | ||
XM_006255625.2 | XP_006255687.1 | 5 | 1818 | |||
XM_006255626.2 | XP_006255688.1 | 6 | 2010 | |||
XM_006255627.2 | XP_006255689.1 | 4 | 1700 | |||
GenBank mRNA | CO404237.1 | - | 1 | 33 | ||
EV769154.1 | - | 3 | 504 | |||
X76988.1 | - | 655 |