Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLLT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195626.1 | NP_001182555.1 | 8 | 722 | ||
NM_001324297.1 | NP_001311226.1 | 10 | 1419 | |||
NM_004641.3 | NP_004632.1 | 9 | 1071 | |||
NR_136736.1 | 10 | 1249 | ||||
GenBank mRNA | AB209755.1 | - | 10 | 955 | ||
AY598745.1 | - | 8 | 722 | |||
BC129946.1 | - | 9 | 765 | |||
BX648210.1 | - | 2 | 98 | |||
U13948.1 | - | 9 | 905 |