Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031613.1 | NP_113801.1 | 3 | 368 | ||
XM_006243407.2 | XP_006243469.1 | 4 | 828 | |||
XM_006243408.2 | XP_006243470.1 | 4 | 4030 | |||
XM_008766331.1 | XP_008764553.1 | 4 | 420 | |||
GenBank mRNA | U59240.1 | - | 3 | 368 |