Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031640.1 | NP_113828.1 | 4 | 630 | ||
XM_008765417.1 | XP_008763639.1 | 3 | 731 | |||
GenBank mRNA | AF097723.1 | - | 4 | 630 | ||
AF131077.1 | - | 3 | 572 | |||
BC070884.1 | - | 3 | 591 | 7098277 | ||
FQ210652.1 | - | 2 | 370 |