Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031704.2 | NP_113892.2 | 7, 8 | 735 | ||
XM_006230997.2 | XP_006231059.1 | 6, 7 | 1278 | |||
GenBank mRNA | BC085724.1 | - | 6, 7 | 1061 | ||
BC127489.1 | - | 7, 8 | 735 | 7376571 | ||
L20822.1 | - | 6, 7 | 565 | |||
U87971.1 | - | 7, 8 | 735 |