Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033860.1 | NP_001029032.1 | 2 | 272 | ||
NM_001033861.1 | NP_001029033.1 | 2 | 272 | |||
NM_001033862.1 | NP_001029034.1 | 2 | 272 | |||
NM_031755.2 | NP_113943.1 | 2 | 272 | |||
GenBank mRNA | AJ277104.1 | - | 2 | 234 | ||
AJ277105.1 | - | 2 | 234 | |||
J04963.1 | - | 2 | 279 | |||
M92848.1 | - | 2 | 272 | |||
X91137.1 | - | 2 | 234 |