Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256560.1 | NP_001243489.1 | 1 | 213 | ||
NM_004944.3 | NP_004935.1 | 1 | 213 | |||
GenBank mRNA | AF047354.1 | - | 1 | 186 | ||
AK131323.1 | - | 4 | 708 | |||
AK301263.1 | - | 1 | 179 | |||
AK313303.1 | - | 1 | 179 | |||
BC015831.1 | - | 1 | 183 | 4723725 | ||
BI521582.1 | - | 1 | 423 | |||
BI834035.1 | - | 3 | 601 | |||
U56814.1 | - | 1 | 140 |