Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031728.1 | NP_113916.1 | 10 | 923 | ||
XM_006243491.2 | XP_006243553.1 | 10 | 1060 | |||
XM_006243492.2 | XP_006243554.1 | 10 | 1121 | |||
XM_006243493.2 | XP_006243555.1 | 10 | 1060 | |||
XM_006243494.2 | XP_006243556.1 | 10 | 1060 | |||
XM_006243495.2 | XP_006243557.1 | 10 | 1060 | |||
XM_006243496.2 | XP_006243558.1 | 10 | 1061 | |||
XM_006243497.2 | XP_006243559.1 | 10 | 1060 | |||
XM_008766455.1 | XP_008764677.1 | 10 | 1016 | |||
XM_008766456.1 | XP_008764678.1 | 10 | 1060 | |||
GenBank mRNA | X68877.1 | - | 10 | 923 | ||
X68878.1 | - | 10 | 923 |