Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031798.1 | NP_113986.1 | 4, 5 | 1159 | ||
XM_006254752.2 | XP_006254814.1 | 4, 5 | 1090 | |||
GenBank mRNA | AF051561.1 | - | 4, 5 | 1159 | ||
AF061084.1 | - | 2, 3 | 183 | |||
AF071863.1 | - | 4, 5 | 442 |