Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vegfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110335.1 | NP_001103805.1 | 3 | 510 | ||
NM_001287107.1 | NP_001274036.1 | 6 | 1455 | |||
NM_001287111.1 | NP_001274040.1 | 6 | 1455 | |||
NM_001287112.1 | NP_001274041.1 | 6 | 1455 | |||
NM_031836.3 | NP_114024.2 | 6 | 1455 | |||
GenBank mRNA | AF062644.1 | - | 3 | 106 | ||
AF215725.1 | - | 6 | 441 | |||
AF222779.1 | - | 6 | 441 | |||
AF261751.1 | - | 3 | 110 | |||
AY033508.1 | - | 6 | 452 | |||
CK357826.1 | - | 3 | 510 | |||
CK364938.1 | - | 3 | 300 | |||
EU040285.1 | - | 3 | 100 | |||
L20913.1 | - | 4 | 325 |