- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Calb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031984.2 | NP_114190.1 | 2, 3 | 270 | ||
GenBank mRNA | BC081764.1 | - | 2, 3 | 253 | 7110962 | |
CK479935.1 | - | 2, 3 | 271 | |||
FQ214326.1 | - | 2, 3 | 287 | |||
X04280.1 | - | 2, 3 | 285 |