Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HBB SIRNA Assays ›
Gene Symbol:
Hbb
Gene Name:
hemoglobin, beta
Gene Aliases:
-
Chromosome Location:
Chr.1: 168971269 - 168972680 on Build Rnor_6.0
UniGene ID:
Rn.202945
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
54467
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hbb
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033234.1
NP_150237.1
2
177
GenBank mRNA
BC058448.1
-
2
186
6921704
FQ209466.1
-
2
182
FQ209544.1
-
2
182
FQ209725.1
-
2
184
FQ209747.1
-
2
182
FQ209754.1
-
2
182
FQ209760.1
-
2
182
FQ209781.1
-
2
182
FQ209789.1
-
2
182
FQ209796.1
-
2
182
FQ209806.1
-
2
184
FQ209822.1
-
182
FQ209826.1
-
2
191
FQ209841.1
-
2
184
FQ209870.1
-
2
184
FQ209871.1
-
2
182
FQ209903.1
-
2
182
FQ209929.1
-
2
182
FQ209937.1
-
2
182
FQ209961.1
-
2
181
FQ209989.1
-
2
182
FQ209996.1
-
2
182
FQ210038.1
-
2
184
FQ210054.1
-
2
182
FQ210058.1
-
2
182
FQ210090.1
-
2
184
FQ210123.1
-
2
182
FQ210127.1
-
2
182
FQ210137.1
-
2
182
FQ210175.1
-
2
180
FQ210181.1
-
2
184
FQ210200.1
-
2
182
FQ210211.1
-
2
182
FQ210216.1
-
2
181
FQ210218.1
-
2
182
FQ210280.1
-
2
205
FQ210343.1
-
2
182
FQ210348.1
-
2
182
FQ210353.1
-
2
182
FQ210373.1
-
2
182
FQ210415.1
-
2
182
FQ210449.1
-
2
182
FQ210466.1
-
2
182
FQ210482.1
-
2
182
FQ210486.1
-
2
184
FQ210495.1
-
2
182
FQ210497.1
-
2
183
FQ210499.1
-
2
182
FQ210511.1
-
2
182
FQ210523.1
-
2
182
FQ210534.1
-
2
182
FQ210602.1
-
2
182
FQ210676.1
-
2
182
FQ210677.1
-
2
182
FQ210735.1
-
2
181
FQ210771.1
-
2
182
FQ210783.1
-
2
174
FQ210789.1
-
2
182
FQ210794.1
-
2
182
FQ210798.1
-
2
182
FQ210806.1
-
2
182
FQ210826.1
-
2
182
FQ210828.1
-
2
179
FQ210829.1
-
2
182
FQ210838.1
-
2
182
FQ210852.1
-
2
182
FQ210875.1
-
2
182
FQ210912.1
-
2
182
FQ210937.1
-
2
182
FQ210949.1
-
2
185
FQ210960.1
-
2
182
FQ210984.1
-
2
182
FQ210985.1
-
2
182
FQ210998.1
-
2
182
FQ211014.1
-
2
182
FQ211016.1
-
2
182
FQ211034.1
-
2
182
FQ211061.1
-
2
181
FQ211065.1
-
2
182
FQ211103.1
-
2
182
FQ211125.1
-
2
184
FQ211133.1
-
2
179
FQ211137.1
-
2
182
FQ211140.1
-
2
182
FQ211142.1
-
2
184
FQ211147.1
-
2
182
FQ211154.1
-
2
182
FQ211182.1
-
2
182
FQ211193.1
-
2
182
FQ211207.1
-
2
184
FQ211241.1
-
2
182
FQ211242.1
-
2
182
FQ211290.1
-
2
182
FQ211314.1
-
2
182
FQ211331.1
-
2
182
FQ211336.1
-
2
182
FQ211338.1
-
2
181
FQ211340.1
-
2
182
FQ211423.1
-
2
184
FQ211443.1
-
2
182
FQ211452.1
-
2
182
FQ211477.1
-
2
182
FQ211497.1
-
2
182
FQ211548.1
-
2
182
FQ211583.1
-
2
182
FQ211590.1
-
2
183
FQ211613.1
-
2
184
FQ211617.1
-
2
182
FQ211626.1
-
2
182
FQ211692.1
-
2
182
FQ211696.1
-
2
182
FQ211790.1
-
2
182
FQ211806.1
-
2
182
FQ211815.1
-
2
182
FQ211829.1
-
2
184
FQ211847.1
-
2
182
FQ211858.1
-
2
182
FQ211864.1
-
2
182
FQ211905.1
-
2
182
FQ211931.1
-
2
182
FQ211999.1
-
2
182
FQ212127.1
-
2
182
FQ212156.1
-
2
180
FQ212234.1
-
2
182
FQ212258.1
-
2
181
FQ212275.1
-
2
182
FQ212287.1
-
2
182
FQ212290.1
-
2
182
FQ212303.1
-
2
182
FQ212341.1
-
2
184
FQ212449.1
-
2
182
FQ213270.1
-
2
182
FQ213352.1
-
2
182
FQ213466.1
-
2
182
FQ213534.1
-
2
182
FQ213582.1
-
661
FQ213796.1
-
2
182
FQ214044.1
-
2
182
FQ214573.1
-
2
182
FQ216995.1
-
2
184
FQ217014.1
-
2
184
FQ217108.1
-
2
182
FQ217151.1
-
2
182
FQ217201.1
-
2
182
FQ217206.1
-
2
182
FQ217586.1
-
2
184
FQ217619.1
-
182
FQ217623.1
-
2
182
FQ217653.1
-
2
182
FQ217685.1
-
2
182
FQ217803.1
-
2
151
FQ218033.1
-
2
181
FQ218042.1
-
2
182
FQ218100.1
-
2
182
FQ218112.1
-
2
182
FQ218118.1
-
2
184
FQ218119.1
-
2
181
FQ218124.1
-
2
186
FQ218145.1
-
2
182
FQ218153.1
-
2
182
FQ218162.1
-
2
182
FQ218163.1
-
2
182
FQ218171.1
-
2
182
FQ218180.1
-
2
182
FQ218182.1
-
2
182
FQ218199.1
-
2
185
FQ218229.1
-
2
185
FQ218253.1
-
2
182
FQ218272.1
-
2
179
FQ218288.1
-
2
182
FQ218461.1
-
2
202
FQ218539.1
-
2
181
FQ218748.1
-
2
182
FQ218761.1
-
2
182
FQ218763.1
-
2
182
FQ218779.1
-
2
180
FQ219114.1
-
2
182
FQ219985.1
-
2
131
FQ220085.1
-
2
182
FQ220253.1
-
2
131
FQ221076.1
-
2
182
FQ221202.1
-
2
184
FQ221246.1
-
2
182
FQ221520.1
-
2
182
FQ221738.1
-
2
181
FQ221956.1
-
2
182
FQ221962.1
-
2
182
FQ222032.1
-
2
182
FQ222179.1
-
2
182
FQ222262.1
-
2
182
FQ222275.1
-
2
182
FQ222291.1
-
2
182
FQ222503.1
-
2
182
FQ222804.1
-
2
182
FQ222942.1
-
2
182
FQ222960.1
-
2
182
FQ223002.1
-
2
182
FQ223079.1
-
2
182
FQ223164.1
-
2
179
FQ223490.1
-
2
182
FQ223631.1
-
2
182
FQ223717.1
-
2
182
FQ223745.1
-
2
184
FQ223805.1
-
2
182
FQ223852.1
-
2
174
FQ223888.1
-
2
182
FQ223889.1
-
2
184
FQ223995.1
-
2
155
FQ224045.1
-
2
184
FQ224088.1
-
2
182
FQ224309.1
-
2
182
FQ224343.1
-
2
182
FQ224371.1
-
2
182
FQ224406.1
-
2
182
FQ224475.1
-
2
182
FQ224682.1
-
2
182
FQ224727.1
-
2
182
FQ224732.1
-
2
182
FQ225124.1
-
2
244
FQ225436.1
-
2
182
FQ225457.1
-
2
182
FQ225469.1
-
2
182
FQ225684.1
-
334
FQ225738.1
-
2
182
FQ225754.1
-
2
181
FQ225816.1
-
2
182
FQ225817.1
-
2
182
FQ226188.1
-
2
180
FQ226198.1
-
183
FQ226536.1
-
2
182
FQ226572.1
-
2
182
FQ226889.1
-
2
182
FQ227120.1
-
2
182
FQ227384.1
-
183
FQ227498.1
-
2
182
FQ227822.1
-
2
182
FQ227875.1
-
2
182
FQ228016.1
-
2
182
FQ228244.1
-
2
182
FQ228281.1
-
2
182
FQ228294.1
-
2
182
FQ228295.1
-
2
182
FQ228296.1
-
2
184
FQ228414.1
-
2
182
FQ228616.1
-
2
182
FQ228638.1
-
2
182
FQ229015.1
-
2
182
FQ229033.1
-
2
182
FQ229489.1
-
2
182
FQ230471.1
-
2
182
FQ230486.1
-
2
180
FQ230503.1
-
2
182
FQ230508.1
-
2
182
FQ230541.1
-
2
182
FQ230717.1
-
2
182
FQ230861.1
-
2
182
FQ230866.1
-
2
182
FQ230889.1
-
2
182
FQ231191.1
-
2
181
FQ231199.1
-
2
182
FQ231208.1
-
2
181
FQ231230.1
-
2
182
FQ231253.1
-
2
182
FQ231284.1
-
2
184
FQ231289.1
-
2
182
FQ232390.1
-
450
FQ232655.1
-
2
214
FQ233311.1
-
2
257
FQ234713.1
-
2
184
FQ234942.1
-
2
180
FQ235254.1
-
818
M17084.1
-
2
134
X16417.1
-
2
177
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: AB364477, AB020013.
Gene Ontology Categories:
Function(s)
Process(es)
glutathione metabolic process
regulation of eIF2 alpha phosphorylation by heme
oxygen transport
hemopoiesis
erythrocyte development
protein heterooligomerization
oxygen transporter activity
iron ion binding
oxygen binding
heme binding
hemoglobin alpha binding
hemoglobin beta binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-b8dk7:80/10.239.160.22:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0