Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: LGI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308275.1 | NP_001295204.1 | 5 | 825 | ||
NM_001308276.1 | NP_001295205.1 | 3 | 681 | |||
NM_005097.3 | NP_005088.1 | 5 | 825 | |||
NR_131777.1 | 3 | 690 | ||||
XM_017016911.1 | XP_016872400.1 | 5 | 758 | |||
XM_017016912.1 | XP_016872401.1 | 3 | 720 | |||
GenBank mRNA | AB209408.1 | - | 1 | 1873 | ||
AF055636.1 | - | 5 | 713 | |||
AF473548.1 | - | 5 | 713 | |||
AK289706.1 | - | 5 | 501 | |||
AK303956.1 | - | 3 | 519 | |||
AK309416.1 | - | 3 | 565 | |||
AY358885.1 | - | 5 | 671 | |||
BC022500.1 | - | 5 | 749 | 4811956 | ||
KU178468.1 | - | 5 | 489 | |||
KU178469.1 | - | 5 | 498 | |||
KU178470.1 | - | 5 | 489 | |||
KU178471.1 | - | 3 | 345 |