Search Thermo Fisher Scientific
Gene Symbol: Scd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031841.1 | NP_114029.1 | 2 | 643 | ||
GenBank mRNA | AB032243.1 | - | 2 | 643 | ||
BC061737.1 | - | 2 | 585 | 5599134 |