Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Src | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031977.1 | NP_114183.1 | 3, 4 | 365 | ||
XM_008762387.1 | XP_008760609.1 | 3, 4 | 374 | |||
XM_008762388.1 | XP_008760610.1 | 3, 4 | 411 | |||
GenBank mRNA | AF157016.1 | - | 3, 4 | 365 |