Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032066.1 | NP_114455.1 | 3 | 281 | ||
XM_006234599.2 | XP_006234661.1 | 3 | 448 | |||
GenBank mRNA | BC061543.1 | - | 3 | 302 | 6888949 | |
FQ213557.1 | - | 3 | 317 | |||
FQ214777.1 | - | 3 | 346 | |||
U81186.1 | - | 3 | 281 |