Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Chn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032083.1 | NP_114472.1 | 5 | 1054 | ||
XM_006234386.2 | XP_006234448.1 | 11 | 1770 | |||
XM_006234388.2 | XP_006234450.1 | 11 | 1010 | |||
XM_006234389.2 | XP_006234451.1 | 11 | 1713 | |||
XM_008761932.1 | XP_008760154.1 | 8 | 1770 | |||
GenBank mRNA | BC094519.1 | - | 5 | 1103 | 7315609 | |
X67250.1 | - | 5 | 1054 |