Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112742.1 | NP_001106213.1 | 4 | 685 | ||
NM_032990.2 | NP_116785.2 | 4 | 685 | |||
XM_006257496.2 | XP_006257558.1 | 5 | 762 | |||
XM_006257498.2 | XP_006257560.1 | 5 | 785 | |||
XM_006257499.2 | XP_006257561.1 | 4 | 814 | |||
XM_006257501.2 | XP_006257563.1 | 5 | 757 | |||
XM_008773512.1 | XP_008771734.1 | 4 | 814 | |||
GenBank mRNA | M36420.1 | - | 4 | 629 | ||
M38062.1 | - | 4 | 629 | |||
M85036.2 | - | 5 | 795 | |||
X54656.1 | - | 4 | 685 |