Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033715.1 | NP_001028887.1 | 5 | 423 | ||
NM_001033716.1 | NP_001028888.1 | 5 | 423 | |||
NM_053295.2 | NP_445747.2 | 5 | 423 | |||
XM_006231698.2 | XP_006231760.1 | 5 | 831 | |||
XM_006231699.2 | XP_006231761.1 | 5 | 625 | |||
GenBank mRNA | AF346597.1 | - | 3 | 104 | ||
AF346598.1 | - | 3 | 104 | |||
BC070882.1 | - | 5 | 322 | |||
BC091239.1 | - | 5 | 423 | 7324026 | ||
DQ186624.1 | - | 5 | 617 | |||
DQ186625.1 | - | 5 | 617 | |||
DQ186626.1 | - | 5 | 617 | |||
DQ186627.1 | - | 5 | 617 | |||
DQ186628.1 | - | 4 | 578 | |||
DQ186629.1 | - | 4 | 578 | |||
X62520.1 | - | 3 | 99 | |||
Y13587.1 | - | 3 | 116 | |||
Y13588.1 | - | 3 | 116 | |||
Y13589.1 | - | 3 | 116 | |||
Y13590.1 | - | 3 | 116 | |||
Y13591.1 | - | 3 | 116 |