Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053341.1 | NP_445793.1 | 1 | 284 | ||
XM_006255311.2 | XP_006255373.1 | 1 | 2549 | |||
XM_006255312.2 | XP_006255374.1 | 2 | 1543 | |||
XM_008772420.1 | XP_008770642.1 | 2 | 1473 | |||
GenBank mRNA | AF032120.1 | - | 1 | 284 | ||
AF089817.1 | - | 2 | 302 | |||
BC061964.1 | - | 2 | 308 | |||
BC070505.1 | - | 2 | 327 | 7097697 |