Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053289.1 | NP_445741.1 | 2, 3 | 143 | ||
GenBank mRNA | FQ232415.1 | - | 2, 3 | 143 | ||
FQ233015.1 | - | 2, 3 | 143 | |||
M55149.1 | - | 2, 3 | 145 | |||
M98049.1 | - | 1, 2 | 93 | |||
S43715.1 | - | 2, 3 | 143 | |||
S77413.1 | - | 2, 3 | 141 |