Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053311.1 | NP_445763.1 | 2, 3 | 480 | ||
XM_006241294.2 | XP_006241356.1 | 2, 3 | 572 | |||
XM_006241295.2 | XP_006241357.1 | 1, 2 | 449 | |||
XM_006241296.2 | XP_006241358.1 | 2, 3 | 572 | |||
GenBank mRNA | FQ212562.1 | - | 2, 3 | 725 | ||
FQ227803.1 | - | 1, 2 | 289 | |||
J03753.1 | - | 2, 3 | 480 |