Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptafr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053321.2 | NP_445773.1 | 2 | 910 | ||
XM_006239063.2 | XP_006239125.1 | 2 | 1235 | |||
XM_006239064.2 | XP_006239126.1 | 2 | 1521 | |||
GenBank mRNA | BC072491.1 | - | 2 | 910 | 7097307 | |
U04740.1 | - | 2 | 931 |
Set Membership: |
G-protein coupled receptor |