Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053347.1 | NP_445799.1 | 2, 3 | 245 | ||
XM_006245849.2 | XP_006245911.1 | 3, 4 | 362 | |||
XM_006245850.2 | XP_006245912.1 | 4, 5 | 394 | |||
XM_006245851.1 | XP_006245913.1 | 3, 4 | 362 | |||
XM_008767443.1 | XP_008765665.1 | 3, 4 | 346 | |||
XM_008767444.1 | XP_008765666.1 | 3, 4 | 346 | |||
XM_008767445.1 | XP_008765667.1 | 3, 4 | 346 | |||
GenBank mRNA | AF240463.1 | - | 2, 3 | 245 | ||
BC081759.1 | - | 2, 3 | 257 | 7190483 |
Gene Symbol: LOC103690118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245690.2 | XP_006245752.1 | 3, 4 | 391 | ||
XM_008767446.1 | XP_008765668.1 | 4, 5 | 432 |