Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052805.2 | NP_434692.2 | 5 | 1045 | ||
GenBank mRNA | AY574254.1 | - | 5 | 1045 | ||
L31621.1 | - | 5 | 1121 | |||
X03440.1 | - | 5 | 1195 |