Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053288.2 | NP_445740.1 | 5, 6 | 583 | ||
GenBank mRNA | FQ209739.1 | - | 5, 6 | 585 | ||
FQ210338.1 | - | 5, 6 | 585 | |||
FQ218200.1 | - | 5, 6 | 588 | |||
FQ218302.1 | - | 5, 6 | 585 | |||
FQ219945.1 | - | 5, 6 | 589 | |||
FQ220896.1 | - | 5, 6 | 589 | |||
FQ221011.1 | - | 5, 6 | 588 | |||
FQ221095.1 | - | 5, 6 | 584 | |||
FQ221803.1 | - | 4, 5 | 379 | |||
FQ221848.1 | - | 5, 6 | 585 | |||
FQ222215.1 | - | 5, 6 | 585 | |||
FQ222231.1 | - | 4, 5 | 396 | |||
FQ222829.1 | - | 5, 6 | 585 | |||
FQ223035.1 | - | 5, 6 | 585 | |||
FQ228353.1 | - | 5, 6 | 499 | |||
FQ228371.1 | - | 5, 6 | 589 | |||
FQ228464.1 | - | 5, 6 | 583 | |||
FQ228466.1 | - | 5, 6 | 585 | |||
FQ228489.1 | - | 5, 6 | 585 | |||
FQ228584.1 | - | 5, 6 | 585 | |||
FQ228800.1 | - | 5, 6 | 585 | |||
FQ229026.1 | - | 5, 6 | 584 | |||
FQ229053.1 | - | 4, 5 | 309 | |||
FQ229515.1 | - | 5, 6 | 503 | |||
FQ229654.1 | - | 5, 6 | 583 | |||
FQ230009.1 | - | 5, 6 | 589 | |||
J00696.1 | - | 5, 6 | 584 | |||
V01216.1 | - | 5, 6 | 583 |