Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053416.2 | NP_445868.2 | 3 | 456 | ||
XM_006234108.2 | XP_006234170.2 | 4 | 794 | |||
XM_008761777.1 | XP_008759999.1 | 4 | 494 | |||
XM_008761778.1 | XP_008760000.1 | 4 | 494 | |||
GenBank mRNA | CO564209.1 | - | 4 | 533 |