Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053428.1 | NP_445880.1 | 2, 3 | 259 | ||
XM_006257592.1 | XP_006257654.1 | 2, 3 | 527 | |||
XM_008773572.1 | XP_008771794.1 | 3, 4 | 435 | |||
XM_008773573.1 | XP_008771795.1 | 3, 4 | 497 | |||
GenBank mRNA | AF271786.1 | - | 2, 3 | 259 |