- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Stau1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109907.1 | NP_001103377.1 | 3, 4 | 493 | ||
NM_053436.2 | NP_445888.1 | 3, 4 | 493 | |||
GenBank mRNA | AF227200.1 | - | 3, 4 | 480 | ||
AF290989.1 | - | 3, 4 | 480 | |||
AJ010200.1 | - | 1, 2 | 174 | |||
BC081755.1 | - | 3, 4 | 451 | |||
BC101858.1 | - | 3, 4 | 489 | 7937343 | ||
DN932990.1 | - | 3, 4 | 522 |